1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
Supplier: Apollo Scientific
Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
PC5766-25G
APOSPC5766-25GEA 89.9 EUR
APOSPC5766-25G
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
Formula: C₇H₁₁F₃N₂O₃S
MW: 260.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210008
CAS Number: 145022-44-2