N-(4-Fluorophenyl)anthranilic acid 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-[(4-Fluorophenyl)amino]benzoic acid
PC5776-1G
PC5776-5G
APOSPC5776-1GEA
43.5
EUR
APOSPC5776-1G
APOSPC5776-5G
N-(4-Fluorophenyl)anthranilic acid 98%
N-(4-Fluorophenyl)anthranilic acid
Formula:
C₁₃H₁₀FNO₂ MW: 231.22 g/mol Melting Pt: 198…200 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01675229 CAS Number: 54-60-4 |