(±)-3-Fluoro-α-methylbenzylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(3-Fluorophenyl)ethanamine
PC5782-1G PC5782-5G
APOSPC5782-1GEA 115 EUR
APOSPC5782-1G APOSPC5782-5G
(±)-3-Fluoro-α-methylbenzylamine 97%
(±)-3-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 145 °C
Density: 1.059 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD04038308
CAS Number: 74788-45-7
UN: 2735
ADR: 8,III