1-Cyclopropyl-2-(2-fluorophenyl)ethanone
Supplier: Apollo Scientific
Warning
Synonyms: Cyclopropyl 2-Fluorobenzyl ketone
PC5806-5G PC5806-1G
APOSPC5806-5GEA 69.6 EUR
APOSPC5806-5G APOSPC5806-1G
1-Cyclopropyl-2-(2-fluorophenyl)ethanone
1-Cyclopropyl-2-(2-fluorophenyl)ethanone
Formula: C₁₁H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD10687164
CAS Number: 150322-73-9