2-Bromo-4-fluoropyridine
Supplier: Apollo Scientific
Danger
PC5846-1G PC5846-500MG
APOSPC5846-1GEA 21.9 EUR
APOSPC5846-1G APOSPC5846-500MG
2-Bromo-4-fluoropyridine
2-Bromo-4-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
Density: 1.699 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260909
CAS Number: 357927-50-5