2-Bromo-4-(trifluoromethyl)phenylacetonitrile
Supplier: Apollo Scientific
Warning
PC5873-1G PC5873-25G PC5873-5G
APOSPC5873-1GEA 52.2 EUR
APOSPC5873-1G APOSPC5873-25G APOSPC5873-5G
2-Bromo-4-(trifluoromethyl)phenylacetonitrile
2-Bromo-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅BrF₃N
MW: 264.04 g/mol
Boiling Pt: 256…266 °C
Density: 1.626 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094308
CAS Number: 474024-36-7