1H,1H,2H,3H-Nonafluorohept-2-en-1-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Nonafluoro-n-butyl)prop-2-enol
PC5906K-1G PC5906K-5G
APOSPC5906K-1GEA 69.8 EUR
APOSPC5906K-1G APOSPC5906K-5G
1H,1H,2H,3H-Nonafluorohept-2-en-1-ol 98%
1H,1H,2H,3H-Nonafluorohept-2-en-1-ol
Formula: C₇H₅F₉O
MW: 276.1 g/mol
Boiling Pt: 78…83 °C/28mmHg
Density: 1.552 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042340
CAS Number: 72016-05-8