Perfluoro(2-butyltetrahydrofuran)

Supplier: Apollo Scientific
Warning

Synonyms: Perfluoro-2-butyltetrahydrofuran

PC5910-10G PC5910-1G PC5910-5G
APOSPC5910-10GEA 434 EUR
APOSPC5910-10G APOSPC5910-1G APOSPC5910-5G
Perfluoro(2-butyltetrahydrofuran)
Perfluoro(2-butyltetrahydrofuran)

97% as C8F16O; contains:Perfluoro(2-propyl-1-oxacyclohexane)

Formula: C₈F₁₆O
MW: 416.06 g/mol
Boiling Pt: 99…107 °C
Melting Pt: –88 °C
Density: 1.77 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014513
CAS Number: 335-36-4
EINECS: 206-389-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR