2-Bromo-5-chloro-3-fluoropyridine
Supplier: Apollo Scientific
Warning
PC5924-1G PC5924-5G
APOSPC5924-1GEA 21.9 EUR
APOSPC5924-1G APOSPC5924-5G
2-Bromo-5-chloro-3-fluoropyridine
2-Bromo-5-chloro-3-fluoropyridine
Formula: C₅H₂BrClFN
Boiling Pt: 77…78 °C/7mmHg
Melting Pt: 28…30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044268
CAS Number: 514797-97-8