4-Cyano-3-fluorobenzoic acid
Supplier: Apollo Scientific
Warning
PC5929-1G PC5929-5G
APOSPC5929-1GEA 21.9 EUR
APOSPC5929-1G APOSPC5929-5G
4-Cyano-3-fluorobenzoic acid
4-Cyano-3-fluorobenzoic acid
Formula: C₈H₄FNO₂
MW: 165.1213 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD03094324
CAS Number: 176508-81-9