2-Fluoro-4-methylbenzaldehyde 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-p-tolylaldehyde, 2-Fluoro-4-tolualdehyde, 2-Fluoro-p-tolualdehyde
PC5933-1G PC5933-5G PC5933-10G
APOSPC5933-1GEA 21.9 EUR
APOSPC5933-1G APOSPC5933-5G APOSPC5933-10G
2-Fluoro-4-methylbenzaldehyde 95%
2-Fluoro-4-methylbenzaldehyde
Formula: C₈H₇FO
Boiling Pt: 91…92 °C/14mmHg
Density: 0.95 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD03094325
CAS Number: 146137-80-6