Ethyl-4,4-difluoroacetoacetate 96%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-3-oxobutyrate
PC5945-100G PC5945-25G PC5945-500G
APOSPC5945-100GEA 75.4 EUR
APOSPC5945-100G APOSPC5945-25G APOSPC5945-500G
Ethyl-4,4-difluoroacetoacetate 96%
Ethyl 4,4-difluoroacetoacetate
Formula: C₆H₈F₂O₃
Boiling Pt: 162 °C
Density: 1.2418 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039277
CAS Number: 352-24-9
EINECS: 206-519-0
UN: 1993
ADR: 3,III