1,1-Difluoroacetylacetone 97%
Supplier: Apollo Scientific
Danger
PC5946-25G PC5946-5G
APOSPC5946-25GEA 553 EUR
APOSPC5946-25G APOSPC5946-5G
1,1-Difluoroacetylacetone 97%
1,1-Difluoroacetylacetone
Formula: C₅H₆F₂O₂
Boiling Pt: 132…133 °C
Storage Temperature: Ambient
MDL Number: MFCD02254117
CAS Number: 41739-23-5
UN: 1993
ADR: 3,II