1-Ethynyl-3,5-bis(trifluoromethyl)benzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3',5'-Bis(trifluoromethyl)phenylacetylene
PC5951-1G PC5951-25G PC5951-5G
APOSPC5951-1GEA 21.9 EUR
APOSPC5951-1G APOSPC5951-25G APOSPC5951-5G
1-Ethynyl-3,5-bis(trifluoromethyl)benzene 99%
1-Ethynyl-3,5-bis(trifluoromethyl)benzene
Formula: C₁₀H₄F₆
MW: 238.13 g/mol
Boiling Pt: 147…148 °C
Density: 1.346 g/cm³ (25 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD01861850
CAS Number: 88444-81-9
UN: 1993
ADR: 3,III