1-Fluoro-4-ethynylbenzene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne
PC5955-1G PC5955-25G PC5955-5G
APOSPC5955-1GEA 21.9 EUR
APOSPC5955-1G APOSPC5955-25G APOSPC5955-5G
1-Fluoro-4-ethynylbenzene 98%
1-Fluoro-4-ethynylbenzene
Formula: C₈H₅F
MW: 120.13 g/mol
Boiling Pt: 55…56 °C/40mmHg
Melting Pt: 27…28 °C
Density: 1.048 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1 II