1-Fluoro-3-ethynylbenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Fluorophenyl)ethyne, 3-Fluorophenylacetylene, (3-Fluorophenyl)acetylene, 1-Ethynyl-3-fluorobenzene
PC5956-100G PC5956-25G PC5956-5G
APOSPC5956-100GEA 209 EUR
APOSPC5956-100G APOSPC5956-25G APOSPC5956-5G
1-Fluoro-3-ethynylbenzene 98%
1-Fluoro-3-ethynylbenzene
Formula: C₈H₅F
Boiling Pt: 132 °C
Density: 1.039 g/cm³ (25 °C)
Flash Pt: 32.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03093798
CAS Number: 2561-17-3
UN: 1993
ADR: 3,III