2,5-Bis(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,5-Bis(trifluoromethyl)benzoylchloride, 2,5-Bis(trifluoromethyl)benzene-1-carbonyl chloride
PC5976-1G PC5976-5G
APOSPC5976-1GEA 23.2 EUR
APOSPC5976-1G APOSPC5976-5G
2,5-Bis(trifluoromethyl)benzoyl chloride 97%
2,5-Bis(trifluoromethyl)benzoyl chloride
Formula: C₉H₃ClF₆O
Boiling Pt: 185 °C
Density: 1.528 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00074997
CAS Number: 393-82-8
UN: 3265
ADR: 8,II