2-Chloro-4-fluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC5988-25G PC5988-100G PC5988-5G
APOSPC5988-25GEA 56.7 EUR
APOSPC5988-25G APOSPC5988-100G APOSPC5988-5G
2-Chloro-4-fluoropyridine 98%
2-Chloro-4-fluoropyridine
Formula: C₅H₃ClFN
Boiling Pt: 140 °C
Density: 1.456 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD04039345
CAS Number: 34941-91-8
UN: 1993
ADR: 3,III