1H,1H,2H,2H-Heneicosafluoro-1-dodecanol 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1H,1H,2H,2H-Perfluoro-1-dodecanol, 1H,1H,2H,2H-Perfluorododecan-1-ol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecanol
PC5996-250MG PC5996-1G
APOSPC5996-250MGEA 36.4 EUR
APOSPC5996-250MG APOSPC5996-1G
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol 97%
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol
Formula: C₁₂H₅F₂₁O
MW: 564.14 g/mol
Boiling Pt: 145 °C/10mmHg
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00039545
CAS Number: 865-86-1
EINECS: 212-748-7
UN: 1325
ADR: 4.1,II