2-(Trifluoromethyl)-1,8-naphthyridine-3-carboxylic acid 95+%
Supplier: Apollo Scientific
Warning
PC5999-1G PC5999-250MG
APOSPC5999-1GEA 159 EUR
APOSPC5999-1G APOSPC5999-250MG
2-(Trifluoromethyl)-1,8-naphthyridine-3-carboxylic acid 95+%
2-(Trifluoromethyl)-1,8-naphthyridine-3-carboxylic acid
Formula: C₁₀H₅F₃N₂O₂
MW: 242.16 g/mol
Melting Pt: 309…310 °C
Storage Temperature: Ambient
MDL Number: MFCD00202890
CAS Number: 241154-08-5