1H,1H,2H-Perfluoro-1-dodecene 97%
Supplier: Apollo Scientific
Warning
PC6003-1G PC6003-25G PC6003-5G
APOSPC6003-1GEA 26.1 EUR
APOSPC6003-1G APOSPC6003-25G APOSPC6003-5G
1H,1H,2H-Perfluoro-1-dodecene 97%
1H,1H,2H-Perfluoro-1-dodecene
Formula: C₁₂H₃F₂₁
MW: 546.12 g/mol
Boiling Pt: 71…72 °C/14mmHg
Density: 1.711 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042346
CAS Number: 30389-25-4
EINECS: 250-173-3