1-Iodoperfluorododecane 97%

Supplier: Apollo Scientific
Warning

Synonyms: Pentacosafluoro-1-iodododecane, Perfluorododecyl iodide

PC6003D-25G
APOSPC6003D-25GEA 319 EUR
APOSPC6003D-25G
1-Iodoperfluorododecane 97%
1-Iodoperfluorododecane
Formula: C₁₂F₂₅I
MW: 745.99 g/mol
Boiling Pt: 108…110 °C/18mmHg
Melting Pt: 95…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001066
CAS Number: 307-60-8
EINECS: 206-205-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR