1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluoro-12-iodododecane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1H,1H,2H,2H-Perfluorododecyl iodide, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Henicosafluoro-12-iodododecane, 1H,1H,2H,2H-Perfluoro-1-iodododecane
PC6003M-25G PC6003M-5G
APOSPC6003M-25GEA 340 EUR
APOSPC6003M-25G APOSPC6003M-5G
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluoro-12-iodododecane 97%
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluoro-12-iodododecane
Formula: C₁₂H₄F₂₁I
MW: 674.03 g/mol
Boiling Pt: 108 °C/10mmHg
Melting Pt: 82…83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039412
CAS Number: 2043-54-1
EINECS: 218-054-0