3-(1,1,2,2-Tetrafluoroethoxy)propene 98%
Supplier: Apollo Scientific
Danger
PC6007-25G PC6007-5G PC6007-1G
APOSPC6007-25GEA 836 EUR
APOSPC6007-25G APOSPC6007-5G APOSPC6007-1G
3-(1,1,2,2-Tetrafluoroethoxy)propene 98%
3-(1,1,2,2-Tetrafluoroethoxy)propene
Formula: C₅H₆F₄O
MW: 158.1 g/mol
Boiling Pt: 76…78 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00043822
CAS Number: 1428-33-7
UN: 3271
ADR: 3 II