Ethyl-4-chloro-2-trifluoromethylpyrimidine-5-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4-chloro-2-trifluoromethylpyrimidine-5-carboxylate
PC6023-1G
APOSPC6023-1GEA 65.4 EUR
APOSPC6023-1G
Ethyl-4-chloro-2-trifluoromethylpyrimidine-5-carboxylate
Ethyl-4-chloro-2-trifluoromethylpyrimidine-5-carboxylate
Formula: C₈H₆ClF₃N₂O₂
MW: 254.5937 g/mol
Boiling Pt: 95 °C/0.3mmHg
Melting Pt: 36…42 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00173897
CAS Number: 720-01-4