N-Ethyl-4-(trifluoromethoxy)aniline 97%
Supplier: Apollo Scientific
|
Warning
|
PC6041-1G
PC6041-100MG
PC6041-250MG
APOSPC6041-1GEA
118
EUR
APOSPC6041-1G
APOSPC6041-100MG
APOSPC6041-250MG
N-Ethyl-4-(trifluoromethoxy)aniline 97%
N-Ethyl-4-(trifluoromethoxy)aniline
Formula:
C₉H₁₀F₃NO Boiling Pt: 42…44 °C/0.2mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00203992 CAS Number: 175278-20-3 |