5-Chloro-3-fluoropyridine-2-carbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-3-fluoropicolinonitrile
PC6044-1G PC6044-5G PC6044-250MG
APOSPC6044-1GEA 137 EUR
APOSPC6044-1G APOSPC6044-5G APOSPC6044-250MG
5-Chloro-3-fluoropyridine-2-carbonitrile
5-Chloro-3-fluoropyridine-2-carbonitrile
Formula: C₆H₂ClFN₂
MW: 156.55 g/mol
Melting Pt: 72…73 °C
Storage Temperature: Ambient
MDL Number: MFCD07375055
CAS Number: 207994-11-4
UN: 3439
ADR: 6.1,III