5-Bromo-2-nitro-4-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
PC6083-1G PC6083-25G PC6083-5G
APOSPC6083-1GEA 69.6 EUR
APOSPC6083-1G APOSPC6083-25G APOSPC6083-5G
5-Bromo-2-nitro-4-(trifluoromethoxy)aniline 98%
5-Bromo-2-nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₄BrF₃N₂O₃
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD12756436
CAS Number: 755030-18-3