1H,1H,2H,2H-Perfluorohexyl bromide 95%
Supplier: Apollo Scientific
Synonyms: 1-Bromo-3,3,4,4,5,5,6,6,6-nonafluorohexane
PC6085Y-100G PC6085Y-25G
APOSPC6085Y-100GEA 0 EUR
APOSPC6085Y-100G APOSPC6085Y-25G
1H,1H,2H,2H-Perfluorohexyl bromide 95%
1H,1H,2H,2H-Perfluorohexyl bromide
Formula: C₆H₄BrF₉
Boiling Pt: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00077576
CAS Number: 38436-14-5