3-(Perfluoro-n-hexyl)prop-2-en-1-ol 98%
Supplier: Apollo Scientific
Warning
PC6088J-1G PC6088J-5G
APOSPC6088J-1GEA 72.5 EUR
APOSPC6088J-1G APOSPC6088J-5G
3-(Perfluoro-n-hexyl)prop-2-en-1-ol 98%
3-(Perfluoro-n-hexyl)prop-2-en-1-ol
Formula: C₉H₅F₁₃O
MW: 376.12 g/mol
Boiling Pt: 87…89 °C/10mmHg
Density: 1.65 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042355
CAS Number: 38550-47-9