3-(Perfluoro-n-hexyl)-1,2-propenoxide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane, 3-Perfluorohexyl-1,2-epoxypropane
PC6088P-25G
APOSPC6088P-25GEA 161 EUR
APOSPC6088P-25G
3-(Perfluoro-n-hexyl)-1,2-propenoxide 97%
3-(Perfluoro-n-hexyl)-1,2-propenoxide
Formula: C₉H₅F₁₃O
MW: 376.12 g/mol
Boiling Pt: 80 °C/41mmHg
Density: 1.645 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00042356
CAS Number: 38565-52-5
EINECS: 254-004-4