2-(Trifluoromethyl)undecafluoropentane 97%
Supplier: Apollo Scientific
Warning
Synonyms: Tetradecafluoro-2-methylpentane, Perfluoro-2-methylpentane, Perfluoroisohexane
PC6122L-100G PC6122L-500G
APOSPC6122L-100GEA 118 EUR
APOSPC6122L-100G APOSPC6122L-500G
2-(Trifluoromethyl)undecafluoropentane 97%
2-(Trifluoromethyl)undecafluoropentane
Formula: C₆F₁₄
MW: 338.04 g/mol
Boiling Pt: 58.2 °C
Melting Pt: < –150 °C
Density: 1.723 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00059905
CAS Number: 355-04-4
EINECS: 206-575-6