Nonafluoro-2-(trifluoromethyl)-2-pentene 98%
Supplier: Apollo Scientific
Danger
PC6122P-100G PC6122P-1G PC6122P-25G
APOSPC6122P-100GEA 266 EUR
APOSPC6122P-100G APOSPC6122P-1G APOSPC6122P-25G
Nonafluoro-2-(trifluoromethyl)-2-pentene 98%
Nonafluoro-2-(trifluoromethyl)-2-pentene
Formula: C₆F₁₂
Boiling Pt: 48…54 °C
Density: 1.622 g/cm³ (25 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00015724
CAS Number: 1584-03-8
EINECS: 216-436-1
UN: 1993
ADR: 3,II