1H,1H,2H,3H,3H-Tridecafluoro-1,2-nonandiol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 1H,1H,2H,3H,3H-Perfluorononane-1,2-diol, 4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluoro-1,2-nonandiol, 1H,1H,2H,3H,3H-Perfluoro-1,2-nonandiol
PC6123X-1G PC6123X-5G PC6123X-25G
APOSPC6123X-1GEA 72.5 EUR
APOSPC6123X-1G APOSPC6123X-5G APOSPC6123X-25G
1H,1H,2H,3H,3H-Tridecafluoro-1,2-nonandiol 96%
1H,1H,2H,3H,3H-Tridecafluoro-1,2-nonandiol
Formula: C₉H₇F₁₃O₂
MW: 394.13 g/mol
Boiling Pt: 120…130 °C
Melting Pt: 63…64 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00042262
CAS Number: 107650-06-6