1H,1H,2H,2H-Tridecafluoro-1-octanol 97+%
Supplier: Apollo Scientific
Warning
PC6147-100G PC6147-250G PC6147-25G
APOSPC6147-100GEA 61.5 EUR
APOSPC6147-100G APOSPC6147-250G APOSPC6147-25G
1H,1H,2H,2H-Tridecafluoro-1-octanol 97+%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Formula: C₈H₅F₁₃O
Boiling Pt: 91…93 °C/36mmHg
Density: 1.678 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1