2,6-Dichloro-4-(trifluoromethyl)nicotinic acid
Supplier: Apollo Scientific
Warning
PC6153-1G PC6153-500MG
APOSPC6153-1GEA 295 EUR
APOSPC6153-1G APOSPC6153-500MG
2,6-Dichloro-4-(trifluoromethyl)nicotinic acid
2,6-Dichloro-4-(trifluoromethyl)nicotinic acid
Formula: C₇H₂Cl₂F₃NO₂
MW: 260 g/mol
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD06208180
CAS Number: 503437-19-2