2-(Trifluoromethyl)imidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC6159-1G PC6159-500MG
APOSPC6159-1GEA 270 EUR
APOSPC6159-1G APOSPC6159-500MG
2-(Trifluoromethyl)imidazo[1,2-a]pyridine
2-(Trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111687
CAS Number: 73221-12-2