1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, Acrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester
PC6166-25G PC6166-5G PC6166-100G
APOSPC6166-25GEA 36.4 EUR
APOSPC6166-25G APOSPC6166-5G APOSPC6166-100G
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate 97%
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate
Formula: C₁₁H₇F₁₃O₂
MW: 418.154 g/mol
Boiling Pt: 76…80 °C/8mmHg
Density: 1.554 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042351
CAS Number: 17527-29-6
EINECS: 241-527-8