1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Supplier: Apollo Scientific
Danger
Synonyms: Chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl)silane
PC6167E-1G PC6167E-25G PC6167E-5G
APOSPC6167E-1GEA 21.9 EUR
APOSPC6167E-1G APOSPC6167E-25G APOSPC6167E-5G
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Formula: C₁₀H₁₀ClF₁₃Si
MW: 440.71 g/mol
Boiling Pt: 189…191 °C
Density: 1.461 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00042359
CAS Number: 102488-47-1
UN: 2986
ADR: 8+3,II