1H,1H,2H,2H-Perfluoro-1-iodooctane 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 1-Iodo-3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane
PC6172-100G
PC6172-25G
PC6172-5G
APOSPC6172-100GEA
161
EUR
APOSPC6172-100G
APOSPC6172-25G
APOSPC6172-5G
1H,1H,2H,2H-Perfluoro-1-iodooctane 97%
1H,1H,2H,2H-Perfluoro-1-iodooctane
Formula:
C₈H₄F₁₃I Boiling Pt: 178…180 °C Melting Pt: 20…21 °C Density: 1.934 g/cm³ (25 °C) Flash Pt: >150 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039410 CAS Number: 2043-57-4 EINECS: 218-056-1 |