4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Supplier: Apollo Scientific
Warning
PC6174-1G PC6174-250MG
APOSPC6174-1GEA 299 EUR
APOSPC6174-1G APOSPC6174-250MG
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formula: C₁₃H₈F₃NO₃
MW: 283.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703523
CAS Number: 1086379-75-0