1H,1H,2H,2H-Perfluorooctyltrichlorosilane 97%
Supplier: Apollo Scientific
Danger
Synonyms: Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane
PC6179-5G PC6179-25G
APOSPC6179-5GEA 60.9 EUR
APOSPC6179-5G APOSPC6179-25G
1H,1H,2H,2H-Perfluorooctyltrichlorosilane 97%
1H,1H,2H,2H-Perfluorooctyltrichlorosilane
Formula: C₈H₄Cl₃F₁₃Si
MW: 481.55 g/mol
Boiling Pt: 84…85 °C/17mmHg
Density: 1.638 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00042363
CAS Number: 78560-45-9
EINECS: 278-947-6
UN: 2987
ADR: 8 II