2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1H,1H-Nonafluoro-1-pentanol, 1H,1H-Perfluoropentan-1-ol
PC6186-1G PC6186-25G PC6186-5G
APOSPC6186-1GEA 82.8 EUR
APOSPC6186-1G APOSPC6186-25G APOSPC6186-5G
2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol 97%
2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol
Formula: C₅H₃F₉O
MW: 250.06 g/mol
Boiling Pt: 110…111 °C
Density: 1.664 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236714
CAS Number: 355-28-2