Perfluorotriamylamine 85%

Supplier: Apollo Scientific
Warning

Synonyms: Perfluorotripentylamine

PC6220L-25G PC6220L-50G
APOSPC6220L-25GEA 51 EUR
APOSPC6220L-25G APOSPC6220L-50G
Perfluorotriamylamine 85%
Perfluorotriamylamine
Formula: C₁₅F₃₃N
MW: 821.12 g/mol
Boiling Pt: 210…220 °C
Density: 1.94 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042367
CAS Number: 338-84-1
EINECS: 206-421-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR