6-(Trifluoromethyl)anthranilic acid 95+%
Supplier: Apollo Scientific
Warning
PC6221-1G PC6221-5G
APOSPC6221-1GEA 127 EUR
APOSPC6221-1G APOSPC6221-5G
6-(Trifluoromethyl)anthranilic acid 95+%
6-(Trifluoromethyl)anthranilic acid
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD01569538
CAS Number: 314-46-5