N,N-Bis(trifluoromethanesulphonyl)aniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)
PC6227-25G PC6227-5G PC6227-100G
APOSPC6227-25GEA 24.8 EUR
APOSPC6227-25G APOSPC6227-5G APOSPC6227-100G
N,N-Bis(trifluoromethanesulphonyl)aniline 98%
N,N-Bis(trifluoromethanesulphonyl)aniline
Formula: C₈H₅F₆NO₄S₂
MW: 357.25 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00000404
CAS Number: 37595-74-7