1,1,1-Trifluoro-3-phenylacetone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one
PC6229M-1G PC6229M-25G PC6229M-5G
APOSPC6229M-1GEA 43.5 EUR
APOSPC6229M-1G APOSPC6229M-25G APOSPC6229M-5G
1,1,1-Trifluoro-3-phenylacetone 98%
1,1,1-Trifluoro-3-phenylacetone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 51…52 °C/2mmHg
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00068171
CAS Number: 350-92-5
UN: 1224
ADR: 3,III