Potassium fluoride dihydrate 98%
Supplier: Apollo Scientific
|
Danger
|
PC6267R-250G
PC6267R-50G
APOSPC6267R-250GEA
32.2
EUR
APOSPC6267R-250G
APOSPC6267R-50G
Potassium fluoride dihydrate 98%
Potassium fluoride dihydrate
Formula:
FK·2H₂O Boiling Pt: 156 °C Melting Pt: 41 °C Density: 2.454 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00149929 CAS Number: 13455-21-5 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III |