Potassium hexafluoroantimonate 99%
Supplier: Apollo Scientific
Synonyms: Potassium hexafluoroantimonate(V)
PC6279-25G PC6279-100G PC6279-5G
APOSPC6279-25GEA 111 EUR
APOSPC6279-25G APOSPC6279-100G APOSPC6279-5G
Potassium hexafluoroantimonate 99%
Potassium hexafluoroantimonate
Formula: F₆KSb
MW: 274.85 g/mol
Boiling Pt: 1505 °C
Melting Pt: 846 °C
Storage Temperature: Ambient
MDL Number: MFCD00043080
CAS Number: 16893-92-8
UN: 1549
ADR: 6.1,III