N-[1-(4-Fluorophenyl)ethyl]-N-methylamine hydrochloride 98%
Supplier: Apollo Scientific
Synonyms: alpha,N-Dimethyl-4-fluorobenzylamine
PC6293-1G PC6293-250MG
APOSPC6293-1GEA 231 EUR
APOSPC6293-1G APOSPC6293-250MG
N-[1-(4-Fluorophenyl)ethyl]-N-methylamine hydrochloride 98%
N-[1-(4-Fluorophenyl)ethyl]-N-methylamine hydrochloride
Formula: C₉H₁₃ClFN
MW: 189.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07838464
CAS Number: 574731-02-5